Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sufu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025391.2 | NP_001020562.1 | 2 | 416 | ||
NM_015752.3 | NP_056567.2 | 2 | 416 | |||
GenBank mRNA | AF134893.1 | - | 2 | 356 | ||
AJ131692.1 | - | 2 | 417 | |||
AJ308625.1 | - | 2 | 95 | |||
AJ308626.1 | - | 2 | 350 | |||
AK016976.1 | - | 2 | 395 | |||
AK047603.1 | - | 2 | 286 | |||
AK156410.1 | - | 2 | 378 | |||
AK208852.1 | - | 1 | 16 | |||
BC048168.1 | - | 2 | 389 | 6821477 | ||
BC056997.1 | - | 2 | 389 | 6821477 |