Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Carf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285463.1 | NP_001272392.1 | 13 | 1531 | ||
NM_001285473.1 | NP_001272402.1 | 9 | 1121 | |||
NM_139150.5 | NP_631889.1 | 11 | 1404 | |||
GenBank mRNA | AB221600.1 | - | 10 | 1187 | ||
AF454947.1 | - | 11 | 1373 | |||
AK044738.1 | - | 13 | 1532 | |||
BC138644.1 | - | 10 | 1310 | 8861664 | ||
BC138645.1 | - | 10 | 1310 | 8861665 | ||
BC145350.1 | - | 10 | 1306 | |||
EU170502.1 | - | 9 | 1102 | |||
EU170503.1 | - | 7 | 837 |