siRNA Details
Gene Symbol: Ccdc108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039495.1 | NP_001034584.1 | 11 | 1978 | ||
GenBank mRNA | AK038623.1 | - | 8 | 1889 | ||
AK046272.1 | - | 10 | 1789 | |||
AK133278.1 | - | 11 | 1979 | |||
BC150939.1 | - | 11 | 1924 | 9087850 |
