Search
Search

Best1| Gene Symbol: Best1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011913.2 | NP_036043.2 | 9 | 1213 | ||
| GenBank mRNA | AK006549.1 | - | 7 | 832 | ||
| AY450427.1 | - | 8 | 1037 | |||
| BC139060.1 | - | 8 | 1028 | 8862082 | ||
| BC139061.1 | - | 8 | 1028 | 8862083 | ||