Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011917.2 | NP_036047.2 | 9 | 799 | ||
GenBank mRNA | AK004800.1 | - | 9 | 774 | ||
AK011711.1 | - | 9 | 797 | |||
AK031247.1 | - | 9 | 842 | |||
AK053643.1 | - | 9 | 792 | |||
AK133654.1 | - | 9 | 781 | |||
AK163899.1 | - | 8 | 852 | |||
AK167019.1 | - | 9 | 819 | |||
AK168037.1 | - | 9 | 845 | |||
BC054743.1 | - | 9 | 799 | 6400974 | ||
D38517.1 | - | 9 | 737 |