Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290986.1 | NP_001277915.1 | 13 | 1876 | ||
NM_001290987.1 | NP_001277916.1 | 12 | 1761 | |||
NM_030234.2 | NP_084510.2 | 13 | 1891 | |||
GenBank mRNA | AK031592.1 | - | 13 | 1817 | ||
AK162181.1 | - | 13 | 1844 | |||
BC094676.1 | - | 12 | 1527 | 30615673 | ||
BC117821.1 | - | 12 | 1700 | |||
BC117822.1 | - | 13 | 1830 | 40092839 |