Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vtcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178594.3 | NP_848709.2 | 3, 4 | 552 | ||
GenBank mRNA | AK143276.1 | - | 3, 4 | 553 | ||
AK145664.1 | - | 3, 4 | 552 | |||
AY280973.1 | - | 3, 4 | 452 | |||
AY322147.1 | - | 3, 4 | 452 | |||
AY346099.1 | - | 3, 4 | 532 | |||
BC032925.1 | - | 3, 4 | 502 | 1477676 |