Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081203.1 | NP_001074672.1 | 5, 6 | 719 | ||
GenBank mRNA | AB188811.1 | - | 4, 5 | 648 | ||
AK077133.1 | - | 5, 6 | 725 | |||
AK164542.1 | - | 5, 6 | 798 | |||
AK165852.1 | - | 4, 5 | 658 | |||
BC021875.1 | - | 4, 5 | 665 | 4212192 | ||
BC023706.1 | - | 1, 2 | 87 | |||
BC106156.1 | - | 5, 6 | 778 | |||
BC138418.1 | - | 5, 6 | 776 | 8861438 |