Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrgprx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034868.3 | NP_001030040.2 | 2 | 141 | ||
GenBank mRNA | AK148121.1 | - | 2 | 72 | ||
AK148153.1 | - | 2 | 147 | |||
BC141225.1 | - | 2 | 64 | 9055837 | ||
CJ131827.1 | - | 2 | 192 |
Set Membership: |
G-protein coupled receptor |