Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkd1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039700.2 | NP_001034789.2 | 18, 19 | 3972 | ||
NM_001286454.1 | NP_001273383.1 | 17, 18 | 3942 | |||
NM_181544.2 | NP_853522.2 | 17, 18 | 3942 | |||
GenBank mRNA | AB290926.1 | - | 17, 18 | 3942 | ||
AY164486.1 | - | 17, 18 | 3942 | |||
DQ382344.1 | - | 17, 18 | 3942 | |||
DQ382345.1 | - | 18, 19 | 3972 | |||
DQ382346.1 | - | 18, 19 | 3954 | |||
DQ382347.1 | - | 9, 10 | 1488 |