Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apol10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177744.4 | NP_808412.1 | 4 | 944 | ||
GenBank mRNA | AK033521.1 | - | 4 | 946 |