Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Axl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190974.1 | NP_001177903.1 | 11 | 1701 | ||
NM_001190975.1 | NP_001177904.1 | 10 | 1437 | |||
NM_009465.4 | NP_033491.2 | 12 | 1728 | |||
GenBank mRNA | AK033237.1 | - | 11 | 1670 | ||
AK155567.1 | - | 13 | 1614 | |||
AK161294.1 | - | 5 | 401 | |||
AK170010.1 | - | 11 | 1675 | |||
AK170897.1 | - | 11 | 1334 | |||
BC008551.1 | - | 7 | 818 | |||
BC046618.1 | - | 12 | 1565 | 6494383 | ||
BC050914.1 | - | 12 | 1536 | 6418252 | ||
BC058230.1 | - | 12 | 1589 | 6313662 | ||
X63535.1 | - | 12 | 1477 |