Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ceacam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039185.1 | NP_001034274.1 | 6 | 1375 | ||
NM_001039186.1 | NP_001034275.1 | 6 | 1375 | |||
NM_001039187.1 | NP_001034276.1 | 4 | 835 | |||
NM_011926.2 | NP_036056.2 | 4 | 835 | |||
GenBank mRNA | AB236328.2 | - | 4 | 744 | ||
AB236329.1 | - | 4 | 744 | |||
AB236330.2 | - | 5 | 1029 | |||
BC016891.1 | - | 6 | 1327 | |||
BC114998.1 | - | 2 | 307 | |||
BC121189.1 | - | 5 | 932 | |||
M77196.1 | - | 6 | 1353 | |||
X15351.1 | - | 6 | 1387 | |||
X53084.1 | - | 4 | 764 | |||
X67278.1 | - | 4 | 847 | |||
X67279.1 | - | 6 | 1387 | |||
X67280.1 | - | 6 | 1304 | |||
X67281.1 | - | 6 | 1304 | |||
X67282.1 | - | 4 | 847 | |||
X67283.1 | - | 4 | 764 |