Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fto | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011936.2 | NP_036066.2 | 6 | 1100 | ||
GenBank mRNA | AJ237917.1 | - | 6 | 1086 | ||
AK016860.1 | - | 3 | 342 | |||
AK040866.1 | - | 6 | 1078 | |||
AK045465.1 | - | 6 | 1077 | |||
AK046935.1 | - | 6 | 1059 | |||
AK049502.1 | - | 6 | 1093 | |||
AK088881.1 | - | 6 | 1086 | |||
AK129437.1 | - | 6 | 1067 | |||
AK161060.1 | - | 6 | 1059 | |||
BC022222.1 | - | 6 | 1051 | 4237261 | ||
BC057008.1 | - | 6 | 1066 | 5708558 |
Gene Symbol: 4831440D22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028531.1 | - | 206 |