Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MAP2K7 SIRNA Assays ›
          Gene Symbol:
          Map2k7
          Gene Name:
          mitogen-activated protein kinase kinase 7
          Gene Aliases:
          5930412N11Rik, JNKK 2, Jnkk2, MAPKK 7, MEK 7, Mapkk7, Mek7, Mkk7, Prkmk7, sek2
          Chromosome Location:
          Chr.8: 4238740 - 4247897 on Build GRCm38
          UniGene ID:
          Mm.3906
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID MSS218515
          Size 20 nmol
          Availability Made To Order
          Catalog # 1320001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Map2k7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001042557.2 NP_001036022.1 10, 11 1287
          NM_001164172.1 NP_001157644.1 10, 11 1287
          NM_001291777.1 NP_001278706.1 9, 10 1239
          NM_001291778.1 NP_001278707.1 9, 10 1130
          NM_001291783.1 NP_001278712.1 9, 10 1130
          NM_011944.3 NP_036074.2 9, 10 1239
          GenBank mRNA AB005654.1 - 10, 11 1162
          AF060943.1 - 9, 10 1130
          AF060944.1 - 9, 10 1150
          AF060945.1 - 9, 10 1150
          AF060946.1 - 10, 11 1198
          AF060947.1 - 10, 11 1198
          AK031137.1 - 9, 10 1148
          AK165184.1 - 9, 10 1156
          AK219049.1 - 3, 4 271
          BC070467.1 - 10, 11 1199 30615302
          U74463.1 - 9, 10 1096
          U74464.1 - 8, 9 985
          U93030.1 - 9, 10 1130

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001025425, XM_006248768, XM_008768985, XM_006248769, AY879265, and others.

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          positive regulation of protein phosphorylation
          protein phosphorylation
          apoptotic process
          response to stress
          response to osmotic stress
          JNK cascade
          activation of JUN kinase activity
          response to heat
          response to UV
          phosphorylation
          response to tumor necrosis factor
          positive regulation of neuron apoptotic process
          stress-activated MAPK cascade
          nucleotide binding
          magnesium ion binding
          protein kinase activity
          protein serine/threonine kinase activity
          MAP kinase kinase activity
          protein tyrosine kinase activity
          protein binding
          ATP binding
          protein C-terminus binding
          JUN kinase kinase activity
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          enzyme binding
          protein kinase binding
          protein phosphatase binding
          mitogen-activated protein kinase kinase kinase binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0