Search
Search

Ddx3y| Gene Symbol: Ddx3y | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012008.2 | NP_036138.1 | 6 | 555 | ||
| GenBank mRNA | AJ007376.1 | - | 6 | 522 | ||
| AK154382.1 | - | 6 | 515 | |||
| BC021453.1 | - | 6 | 520 | 5043470 | ||
| BC051046.1 | - | 6 | 544 | |||