Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g4f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024145.2 | NP_001019316.1 | 14 | 1594 | ||
GenBank mRNA | AB195278.1 | - | 14 | 1594 | ||
AK076378.1 | - | 10 | 1002 | |||
BC039947.1 | - | 14 | 1443 | |||
BC046400.1 | - | 1 | 37 | |||
BC145426.1 | - | 14 | 1546 | 9053984 | ||
BC145427.1 | - | 14 | 1541 | |||
BC145428.1 | - | 14 | 1546 | 9053986 | ||
BC150771.1 | - | 14 | 1546 | 9087682 | ||
DQ904008.1 | - | 14 | 1594 |