Search
Search

Dhrs4| Gene Symbol: Dhrs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037938.2 | NP_001033027.2 | 5 | 643 | ||
| NM_030686.2 | NP_109611.1 | 5 | 643 | |||
| GenBank mRNA | AB045132.1 | - | 5 | 555 | ||
| AK002691.1 | - | 1 | 84 | |||
| AK018788.1 | - | 5 | 526 | |||
| AK146988.1 | - | 5 | 523 | |||
| AW125533.1 | - | 1 | 316 | |||
| BC003484.1 | - | 5 | 478 | 2811468 | ||
| BC054361.1 | - | 5 | 604 | 5008299 | ||