Search
Search

Dhrs4| Gene Symbol: Dhrs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037938.2 | NP_001033027.2 | 2 | 340 | ||
| NM_030686.2 | NP_109611.1 | 2 | 340 | |||
| GenBank mRNA | AB045132.1 | - | 2 | 252 | ||
| AK018788.1 | - | 2 | 223 | |||
| AK146988.1 | - | 2 | 220 | |||
| BC003484.1 | - | 2 | 175 | 2811468 | ||
| BC054361.1 | - | 2 | 301 | 5008299 | ||
| BY291911.1 | - | 2 | 320 | |||