Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cttnbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080285.1 | NP_525024.1 | 23 | 4833 | ||
GenBank mRNA | AK032356.1 | - | 21 | 3214 | ||
AK078315.1 | - | 6 | 636 | |||
AK081137.1 | - | 19 | 3297 | |||
BC068156.1 | - | 23 | 4594 | |||
BC141407.1 | - | 23 | 4829 | 9056020 |