Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heatr5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081179.1 | NP_001074648.1 | 19, 20 | 2952 | ||
GenBank mRNA | AK079542.1 | - | 19, 20 | 2897 | ||
AK146166.1 | - | 19, 20 | 2934 | |||
AK158478.1 | - | 19, 20 | 2938 | |||
AK164172.1 | - | 12, 13 | 1826 | |||
AK220390.1 | - | 16, 17 | 2484 | |||
BC086652.1 | - | 18, 19 | 2888 | 6856641 |