Search
Search

Islr2| Gene Symbol: Islr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161535.1 | NP_001155007.1 | 2 | 795 | ||
| NM_001161536.1 | NP_001155008.1 | 4 | 976 | |||
| NM_001161537.1 | NP_001155009.1 | 3 | 967 | |||
| NM_001161538.1 | NP_001155010.1 | 4 | 744 | |||
| NM_001161539.1 | NP_001155011.1 | 4 | 706 | |||
| NM_001161540.1 | NP_001155012.1 | 3 | 662 | |||
| NM_001161541.1 | NP_001155013.1 | 3 | 553 | |||
| NM_177193.5 | NP_796167.2 | 3 | 786 | |||
| GenBank mRNA | AK047348.1 | - | 1 | 274 | ||
| AK129367.1 | - | 3 | 472 | |||
| BC059068.1 | - | 3 | 639 | 6825665 | ||
| BC096531.1 | - | 2 | 795 | 6826287 | ||
| CD350184.1 | - | 4 | 708 | |||
| EU497918.1 | - | 3 | 603 | |||