Search
Search

Cpa6| Gene Symbol: Cpa6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289497.1 | NP_001276426.1 | 1, 2 | 326 | ||
| NM_177834.4 | NP_808502.2 | 1, 2 | 326 | |||
| GenBank mRNA | AK078883.1 | - | 1, 2 | 327 | ||
| AY773477.1 | - | 1, 2 | 324 | |||