Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gal3st4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033416.2 | NP_001028588.1 | 3 | 2599 | ||
GenBank mRNA | AK132558.1 | - | 3 | 2600 | ||
AK138993.1 | - | 4 | 1264 | |||
AK162468.1 | - | 1 | 824 |
Gene Symbol: 6330418K02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031849.1 | - | 2 | 94 |