Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bend3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199028.2 | NP_950193.1 | 4 | 988 | ||
GenBank mRNA | AK122525.1 | - | 2 | 594 | ||
BC060240.1 | - | 4 | 988 | 6856073 |