Search
Search

Trim46| Gene Symbol: Trim46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039466.1 | NP_001034555.1 | 4, 5 | 833 | ||
| NM_183037.2 | NP_898858.1 | 4, 5 | 936 | |||
| GenBank mRNA | AK132368.1 | - | 4, 5 | 802 | ||
| AK170904.1 | - | 4, 5 | 834 | |||
| AY251388.1 | - | 4, 5 | 816 | |||
| BC065049.1 | - | 4, 5 | 979 | 6418851 | ||
| BC082299.1 | - | 4, 5 | 936 | |||