Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec14l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029937.2 | NP_001025108.1 | 7 | 663 | ||
GenBank mRNA | AK136060.1 | - | 1 | 16 | ||
BC138982.1 | - | 7 | 663 | 8862004 | ||
BC138985.1 | - | 7 | 663 | 8862007 | ||
BC145485.1 | - | 7 | 706 |