Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmo7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201529.2 | NP_963287.2 | 28 | 4946 | ||
GenBank mRNA | AK047836.1 | - | 4 | 1875 | ||
AK129231.1 | - | 25 | 4731 | |||
BC082553.1 | - | 26 | 4961 | |||
BC145011.1 | - | 21 | 3430 | 9053559 | ||
EF071857.1 | - | 28 | 4917 |