Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025885.3 | NP_080161.1 | 2 | 439 | ||
GenBank mRNA | AK005399.1 | - | 2 | 407 | ||
AK010552.1 | - | 2 | 417 | |||
AK160579.1 | - | 2 | 405 | |||
AK168787.1 | - | 2 | 407 | |||
AK209282.1 | - | 2 | 207 | |||
BB611514.1 | - | 2 | 439 | |||
BC044207.1 | - | 2 | 301 | |||
BC044302.1 | - | 2 | 301 | |||
BC055404.1 | - | 2 | 394 | |||
BC082559.1 | - | 2 | 404 | 30606373 |