Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145900.1 | NP_001139372.1 | 5 | 1560 | ||
NM_201646.2 | NP_964008.2 | 4 | 1642 | |||
GenBank mRNA | AI118479.1 | - | 1 | 430 | ||
AK146991.1 | - | 4 | 1474 | |||
BC031195.1 | - | 4 | 1642 | 5059510 |