Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif19a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102615.1 | NP_001096085.1 | 6 | 596 | ||
GenBank mRNA | AK155544.1 | - | 6 | 559 | ||
BC151045.1 | - | 6 | 558 | 9087958 | ||
BC151053.1 | - | 6 | 558 | 9087966 | ||
EF570784.1 | - | 6 | 471 |