Search
Search

Whamm| Gene Symbol: Whamm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004185.3 | NP_001004185.3 | 5 | 1165 | ||
| GenBank mRNA | AK080911.1 | - | 6 | 898 | ||
| AK140311.1 | - | 5 | 1175 | |||
| AK162084.1 | - | 7 | 1170 | |||
| AK220273.1 | - | 5 | 1165 | |||
| BC042749.2 | - | 6 | 1000 | 5322109 | ||
| BC050804.1 | - | 5 | 628 | 6771347 | ||