Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ercc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015769.2 | NP_056584.2 | 11 | 2471 | ||
GenBank mRNA | AF189285.1 | - | 11 | 2471 | ||
BC026792.1 | - | 11 | 2262 | 4911382 | ||
BC051036.1 | - | 10 | 2290 | |||
CO433666.1 | - | 1 | 262 | |||
CX210668.1 | - | 4 | 625 |