Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026504.2 | NP_080780.1 | 3 | 429 | ||
GenBank mRNA | AK007496.1 | - | 3 | 430 | ||
AK009825.1 | - | 3 | 425 | |||
AK014348.1 | - | 3 | 441 | |||
AK045993.1 | - | 3 | 436 | |||
AK050334.1 | - | 3 | 436 | |||
BC052345.1 | - | 3 | 436 | 6585618 |