Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rufy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289774.1 | NP_001276703.1 | 10 | 1310 | ||
NM_001289775.1 | NP_001276704.1 | 10 | 1310 | |||
NM_001289776.1 | NP_001276705.1 | 11 | 1732 | |||
NM_001289777.1 | NP_001276706.1 | 12 | 1557 | |||
NM_027530.3 | NP_081806.1 | 10 | 1678 | |||
GenBank mRNA | AK013185.1 | - | 12 | 1557 | ||
AK018184.1 | - | 10 | 1656 | |||
AK083242.1 | - | 10 | 1613 | |||
AK148795.1 | - | 10 | 974 | |||
AK154941.1 | - | 10 | 1311 | |||
AK160820.1 | - | 10 | 1312 | |||
BC017648.1 | - | 10 | 1067 | 4527858 | ||
BC049127.1 | - | 10 | 1617 | 6821567 | ||
BC058259.1 | - | 11 | 1451 | 6469137 | ||
CT010404.1 | - | 10 | 1051 |