Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018819.4 | NP_061289.1 | 4 | 336 | ||
GenBank mRNA | AF181116.1 | - | 4 | 292 | ||
AK002241.1 | - | 4 | 337 | |||
AK002266.1 | - | 4 | 315 | |||
AK004013.1 | - | 4 | 292 | |||
AK005348.1 | - | 4 | 328 | |||
AK013391.1 | - | 4 | 337 | |||
AK014421.1 | - | 4 | 294 | |||
AK014435.1 | - | 4 | 566 | |||
AK135943.1 | - | 4 | 307 | |||
AK170514.1 | - | 4 | 353 | |||
BC024365.1 | - | 4 | 298 | 4988147 | ||
BC103767.1 | - | 4 | 339 | 30840199 |