Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019878.4 | NP_063931.1 | 5 | 577 | ||
GenBank mRNA | AF022894.1 | - | 5 | 596 | ||
AK033498.1 | - | 5 | 580 | |||
AK087595.1 | - | 5 | 613 | |||
BC024361.1 | - | 4 | 536 | 4990345 | ||
U92076.1 | - | 5 | 522 |