Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204165.1 | NP_001191094.1 | 18 | 1839 | ||
NM_001204166.1 | NP_001191095.1 | 15 | 1731 | |||
NM_019688.2 | NP_062662.1 | 17 | 1785 | |||
GenBank mRNA | AB021132.1 | - | 18 | 1814 | ||
AB037668.1 | - | 9 | 1174 | |||
AB445011.1 | - | 15 | 1723 | |||
AF115480.1 | - | 17 | 1713 | |||
AK004874.1 | - | 9 | 831 | |||
AK220281.1 | - | 18 | 1839 | |||
BC139822.1 | - | 18 | 1771 | 40141811 |