Search
Search

Adam21| Gene Symbol: Adam21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020330.4 | NP_065063.1 | 2 | 1225 | ||
| GenBank mRNA | AF251559.1 | - | 1 | 717 | ||
| AK014827.2 | - | 2 | 1226 | |||
| BC132152.1 | - | 1 | 768 | 40130429 | ||
| BC132344.1 | - | 1 | 768 | 40130621 | ||