Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2l6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019949.2 | NP_064333.2 | 3 | 267 | ||
GenBank mRNA | AF159230.1 | - | 3 | 253 | ||
AK010942.1 | - | 3 | 246 | |||
AK013452.1 | - | 3 | 257 | |||
AK152010.1 | - | 3 | 247 | |||
AK152066.1 | - | 3 | 220 | |||
AK152113.1 | - | 3 | 268 | |||
AK168695.1 | - | 3 | 205 | |||
AK171629.1 | - | 3 | 216 |