Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020593.2 | NP_065618.1 | 8 | 926 | ||
NM_212433.1 | NP_997598.1 | 8 | 926 | |||
GenBank mRNA | AF233226.1 | - | 8 | 865 | ||
AK004544.2 | - | 8 | 878 | |||
AK006392.1 | - | 1 | 38 | |||
AK009024.1 | - | 8 | 865 | |||
AK019550.1 | - | 8 | 866 | |||
AK028048.1 | - | 8 | 888 | |||
AK029594.1 | - | 1 | 45 | |||
AK031185.1 | - | 8 | 863 | |||
AK031428.1 | - | 8 | 886 | |||
AK049870.1 | - | 8 | 884 | |||
AK050195.1 | - | 8 | 878 | |||
AK085686.1 | - | 8 | 881 | |||
AK150097.1 | - | 8 | 878 | |||
AK151130.1 | - | 8 | 877 | |||
AK151857.1 | - | 8 | 877 | |||
AK152780.1 | - | 8 | 877 | |||
AK167075.1 | - | 8 | 864 | |||
BC010212.1 | - | 8 | 827 | |||
BC050758.1 | - | 8 | 879 | |||
BC058253.1 | - | 8 | 883 | 6465107 |