Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021789.2 | NP_068561.1 | 2 | 454 | ||
GenBank mRNA | AF233340.1 | - | 2 | 411 | ||
AK005276.1 | - | 2 | 239 | |||
AK032168.1 | - | 2 | 220 | |||
BC038898.1 | - | 2 | 454 | 5401311 |