Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gapvd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025709.2 | NP_079985.2 | 24 | 4425 | ||
GenBank mRNA | AK014474.1 | - | 9 | 1291 | ||
AK088851.1 | - | 8 | 988 | |||
AK129381.1 | - | 23 | 4020 | |||
BC043715.1 | - | 3 | 389 | |||
BC057164.1 | - | 9 | 1294 | |||
BC060123.1 | - | 24 | 4425 | 6408778 | ||
EF155419.1 | - | 22 | 3843 |