Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025736.2 | NP_080012.1 | 2, 3 | 213 | ||
GenBank mRNA | AF332065.1 | - | 2, 3 | 141 | ||
AF332066.1 | - | 2, 3 | 141 | |||
AK014989.1 | - | 2, 3 | 214 | |||
AK015968.1 | - | 2, 3 | 179 | |||
AK172887.1 | - | 2, 3 | 158 | |||
BC004716.1 | - | 2, 3 | 171 | 3582196 |