Search
Search

Fam187a| Gene Symbol: Fam187a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025766.2 | NP_080042.1 | 1 | 649 | ||
| GenBank mRNA | AK016939.1 | - | 1 | 650 | ||
| AK017119.1 | - | 1 | 650 | |||
| BC116769.1 | - | 1 | 495 | 40126088 | ||
| BC116771.1 | - | 1 | 495 | 40126090 | ||