Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pqlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164420.1 | NP_001157892.1 | 3 | 552 | ||
NM_001164421.1 | NP_001157893.1 | 3 | 484 | |||
NM_001164422.1 | NP_001157894.1 | 3 | 479 | |||
NM_001164423.1 | NP_001157895.1 | 3 | 484 | |||
NM_001164424.1 | NP_001157896.1 | 3 | 479 | |||
NM_025861.3 | NP_080137.2 | 3 | 484 | |||
GenBank mRNA | AK009256.1 | - | 3 | 474 | ||
AK017854.1 | - | 3 | 492 | |||
AK018759.1 | - | 2 | 862 | |||
AK019858.1 | - | 3 | 426 | |||
AK047446.1 | - | 3 | 417 | |||
AK153023.1 | - | 3 | 504 | |||
AK153176.1 | - | 3 | 505 | |||
BC008231.1 | - | 3 | 421 | 2647794 | ||
BC029179.1 | - | 3 | 426 | 3487991 | ||
BC043686.1 | - | 662 | 5135928 | |||
CJ135868.1 | - | 3 | 504 |