Search
Search

Rbm7| Gene Symbol: Rbm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144948.5 | NP_659197.2 | 5 | 703 | ||
| GenBank mRNA | AF458961.1 | - | 5 | 667 | ||
| AK005200.1 | - | 5 | 679 | |||
| AK006755.1 | - | 5 | 668 | |||
| AK008645.1 | - | 5 | 663 | |||
| AK011223.1 | - | 5 | 685 | |||
| AK015662.1 | - | 5 | 705 | |||
| AK145381.1 | - | 4 | 1526 | |||
| AK151056.1 | - | 5 | 682 | |||
| AK153504.1 | - | 6 | 772 | |||
| AK188365.1 | - | 1 | 130 | |||
| AK190328.1 | - | 1 | 130 | |||
| AK192901.1 | - | 1 | 131 | |||
| BC054774.1 | - | 5 | 670 | 6807637 | ||
| BC116931.1 | - | 5 | 652 | 40126250 | ||
| BC119038.1 | - | 5 | 652 | 8733787 | ||