Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmachc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025962.3 | NP_080238.2 | 2 | 255 | ||
GenBank mRNA | AK007725.1 | - | 2 | 190 | ||
AK012761.1 | - | 2 | 192 | |||
AK084194.1 | - | 2 | 206 | |||
BB614283.1 | - | 2 | 256 | |||
BC054756.1 | - | 2 | 194 | 6411797 |