Search
Search

Tbc1d19| Gene Symbol: Tbc1d19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144517.4 | NP_653100.1 | 7, 8 | 606 | ||
| GenBank mRNA | AK012836.1 | - | 7, 8 | 558 | ||
| AK076128.1 | - | 7, 8 | 554 | |||
| AK083084.1 | - | 7, 8 | 577 | |||
| BC020138.1 | - | 7, 8 | 565 | 3983701 | ||