Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113573.1 | NP_001107045.1 | 4 | 619 | ||
NM_001113574.1 | NP_001107046.1 | 4 | 619 | |||
NM_023336.4 | NP_075825.3 | 4 | 779 | |||
GenBank mRNA | AB206708.2 | - | 3 | 430 | ||
AB212272.1 | - | 3 | 428 | |||
AF269193.1 | - | 4 | 567 | |||
AK037435.1 | - | 4 | 601 | |||
AK076472.1 | - | 4 | 550 | |||
AK076924.1 | - | 4 | 698 | |||
AK081471.1 | - | 4 | 600 | |||
AK136007.1 | - | 4 | 578 | |||
AK136429.1 | - | 4 | 550 | |||
AK159827.1 | - | 4 | 605 | |||
AK160752.1 | - | 4 | 578 | |||
AK161164.1 | - | 4 | 575 | |||
AK167573.1 | - | 4 | 577 | |||
AY513269.1 | - | 3 | 445 | |||
BC004832.1 | - | 4 | 564 | |||
BC031536.1 | - | 4 | 593 | 4015879 | ||
CN530570.1 | - | 2 | 194 |